siRNA Details
| Gene Symbol: Rrp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163734.1 | NP_001157206.1 | 12 | 1957 | ||
| NM_028244.2 | NP_082520.2 | 15 | 2273 | |||
| GenBank mRNA | AK129078.1 | - | 12 | 1957 | ||
| BC016569.1 | - | 15 | 2273 | 3157173 | ||

Rrp1b
Genomic Map