siRNA Details
| Gene Symbol: Zfp131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK011832.1 | - | 8 | 1251 | ||
| AK030398.1 | - | 8 | 1450 | |||
| AK088022.1 | - | 1 | 1140 | |||
| AK134257.1 | - | 8 | 1250 | |||
| AK135723.1 | - | 6 | 1336 | |||
| AY092766.1 | - | 6 | 1444 | |||
| BC048839.1 | - | 5 | 933 | |||
| BC057991.1 | - | 7 | 1393 | 5323559 | ||

Zfp131
Genomic Map