siRNA Details
| Gene Symbol: Zfp131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK011832.1 | - | 8 | 1577 | ||
| AK030398.1 | - | 8 | 1776 | |||
| AK088022.1 | - | 1 | 1466 | |||
| AK134257.1 | - | 8 | 1576 | |||
| AK135723.1 | - | 6 | 1662 | |||
| AY092766.1 | - | 6 | 1770 | |||
| BC048839.1 | - | 5 | 1259 | |||
| BC057991.1 | - | 7 | 1719 | 5323559 | ||

Zfp131
Genomic Map