siRNA Details
| Gene Symbol: Plcd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152813.3 | NP_690026.2 | 14 | 2322 | ||
| GenBank mRNA | AK011892.1 | - | 10 | 1253 | ||
| AK046669.1 | - | 14 | 2288 | |||
| AK144950.1 | - | 3395 | ||||
| AK173311.1 | - | 12 | 4280 | |||
| BC031392.1 | - | 14 | 2266 | 4952407 | ||

Plcd3
Genomic Map