siRNA Details
| Gene Symbol: Tmem87b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028248.2 | NP_082524.2 | 8 | 716 | ||
| GenBank mRNA | AK011959.1 | - | 8 | 637 | ||
| AK050203.1 | - | 8 | 708 | |||
| AK164504.1 | - | 6 | 1485 | |||
| AK171811.1 | - | 9 | 632 | |||
| AK172611.1 | - | 9 | 632 | |||
| BC031488.1 | - | 8 | 672 | |||

Tmem87b
Genomic Map