siRNA Details
| Gene Symbol: Hsdl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301367.1 | NP_001288296.1 | 12 | 1723 | ||
| NM_024255.3 | NP_077217.2 | 11 | 1625 | |||
| GenBank mRNA | AJ293845.1 | - | 12 | 1665 | ||
| AK011910.1 | - | 11 | 1573 | |||
| AK085899.1 | - | 11 | 1595 | |||
| AK136773.1 | - | 11 | 1592 | |||
| AK144948.1 | - | 11 | 1578 | |||
| AK145230.1 | - | 11 | 1521 | |||
| AY725196.1 | - | 11 | 1482 | |||
| BC002211.1 | - | 11 | 1516 | 3490303 | ||

Hsdl2
Genomic Map