siRNA Details
Gene Symbol: Rnf219 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026047.4 | NP_080323.3 | 6 | 1601 | ||
GenBank mRNA | BC029231.1 | - | 6 | 1567 | 3156840 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rnf219 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026047.4 | NP_080323.3 | 6 | 1601 | ||
GenBank mRNA | BC029231.1 | - | 6 | 1567 | 3156840 |