siRNA Details
| Gene Symbol: Rnf219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026047.4 | NP_080323.3 | 4 | 437 | ||
| GenBank mRNA | AK013314.1 | - | 4 | 416 | ||
| AK020974.1 | - | 4 | 405 | |||
| BC029231.1 | - | 4 | 403 | 3156840 | ||
| BI986775.1 | - | 4 | 437 | |||

Rnf219
Genomic Map