siRNA Details
| Gene Symbol: Cwf19l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081077.1 | NP_001074546.1 | 3, 4 | 279 | ||
| GenBank mRNA | AK012163.1 | - | 3, 4 | 263 | ||
| AK088543.1 | - | 5, 6 | 1521 | |||
| AK139654.1 | - | 3, 4 | 280 | |||
| BC027553.1 | - | 3, 4 | 264 | |||
| BC037640.1 | - | 3, 4 | 257 | 5096937 | ||

Cwf19l1
Genomic Map