siRNA Details
| Gene Symbol: 2610507B11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002004.2 | NP_001002004.2 | 14 | 1850 | ||
| GenBank mRNA | AK012059.1 | - | 1 | 121 | ||
| AK149719.1 | - | 14 | 1821 | |||
| BC075633.1 | - | 14 | 1810 | 30537398 | ||

2610507B11Rik
Genomic Map