siRNA Details
| Gene Symbol: Fgfbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028263.1 | NP_082539.2 | 2 | 261 | ||
| GenBank mRNA | AK011996.1 | - | 1 | 72 | ||
| AK029452.1 | - | 1 | 64 | |||
| AK167949.1 | - | 2 | 279 | |||
| BC055778.1 | - | 2 | 261 | 6403098 | ||
| DQ503577.1 | - | 1 | 175 | |||

Fgfbp3
Genomic Map