siRNA Details
| Gene Symbol: Fgfbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028263.1 | NP_082539.2 | 2 | 1611 | ||
| GenBank mRNA | AK011996.1 | - | 1 | 1421 | ||
| AK032142.1 | - | 1 | 709 | |||
| AK167949.1 | - | 2 | 1628 | |||
| BC055778.1 | - | 2 | 1611 | 6403098 | ||

Fgfbp3
Genomic Map