siRNA Details
| Gene Symbol: Aldh1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028270.4 | NP_082546.1 | 2 | 306 | ||
| GenBank mRNA | AK012213.1 | - | 2 | 201 | ||
| AK088396.1 | - | 2 | 280 | |||
| AK150992.1 | - | 2 | 208 | |||
| AK151349.1 | - | 2 | 208 | |||
| AK151364.1 | - | 2 | 208 | |||
| AK153416.1 | - | 2 | 208 | |||
| BC020001.1 | - | 2 | 210 | 4207479 | ||
| BC086768.1 | - | 2 | 265 | 6808412 | ||
| CJ158368.1 | - | 2 | 307 | |||

Aldh1b1
Genomic Map