siRNA Details
| Gene Symbol: Spats2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139140.1 | NP_631879.1 | 11 | 1412 | ||
| GenBank mRNA | AB079132.1 | - | 11 | 1412 | ||
| AK159335.1 | - | 11 | 1507 | |||
| AK167788.1 | - | 10 | 1090 | |||
| AK171266.1 | - | 10 | 1168 | |||
| BC023432.1 | - | 9 | 1062 | 5029821 | ||
| BC072579.1 | - | 11 | 1679 | |||

Spats2
Genomic Map