siRNA Details
| Gene Symbol: Pih1d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028300.2 | NP_082576.1 | 5 | 888 | ||
| GenBank mRNA | AK012458.1 | - | 5 | 864 | ||
| AK080744.1 | - | 3 | 508 | |||
| BC019481.1 | - | 5 | 786 | 4190604 | ||
| BC039645.1 | - | 5 | 878 | 5373181 | ||

Pih1d2
Genomic Map