siRNA Details
| Gene Symbol: Hspa12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028306.3 | NP_082582.1 | 4 | 358 | ||
| GenBank mRNA | AK012548.1 | - | 4 | 331 | ||
| AY196790.1 | - | 3 | 238 | |||
| BC011103.1 | - | 4 | 358 | 3990088 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hspa12b| Gene Symbol: Hspa12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028306.3 | NP_082582.1 | 4 | 358 | ||
| GenBank mRNA | AK012548.1 | - | 4 | 331 | ||
| AY196790.1 | - | 3 | 238 | |||
| BC011103.1 | - | 4 | 358 | 3990088 | ||