siRNA Details
| Gene Symbol: Tdrkh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028307.1 | NP_082583.1 | 6 | 835 | ||
| GenBank mRNA | AK012596.1 | - | 6 | 876 | ||
| BC031886.1 | - | 6 | 729 | |||
| BC049363.1 | - | 6 | 835 | 6490411 | ||
| BC057030.1 | - | 6 | 851 | 6833743 | ||

Tdrkh
Genomic Map