siRNA Details
| Gene Symbol: 2700097O09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028314.2 | NP_082590.2 | 5 | 611 | ||
| GenBank mRNA | AK012621.1 | - | 5 | 611 | ||
| AK079348.1 | - | 5 | 610 | |||
| BC056967.1 | - | 5 | 605 | 5694212 | ||
| CX231570.1 | - | 5 | 759 | |||

2700097O09Rik
Genomic Map