siRNA Details
| Gene Symbol: 2700097O09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028314.2 | NP_082590.2 | 7 | 774 | ||
| GenBank mRNA | AK012621.1 | - | 7 | 774 | ||
| AK047049.1 | - | 1 | 1699 | |||
| BC016540.1 | - | 2 | 847 | |||
| BC056967.1 | - | 7 | 768 | 5694212 | ||

2700097O09Rik
Genomic Map