siRNA Details
| Gene Symbol: Skida1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028317.2 | NP_082593.2 | 1 | 1873 | ||
| GenBank mRNA | AK141728.1 | - | 1 | 426 | ||
| BC050860.1 | - | 2 | 1612 | 6331376 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Skida1| Gene Symbol: Skida1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028317.2 | NP_082593.2 | 1 | 1873 | ||
| GenBank mRNA | AK141728.1 | - | 1 | 426 | ||
| BC050860.1 | - | 2 | 1612 | 6331376 | ||