siRNA Details
| Gene Symbol: Hnrnpll | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144802.4 | NP_659051.3 | 6 | 1202 | ||
| GenBank mRNA | AK012866.1 | - | 6 | 918 | ||
| AK034860.1 | - | 6 | 1147 | |||
| AK048900.1 | - | 6 | 991 | |||
| AK146879.1 | - | 6 | 1164 | |||
| AK160943.1 | - | 5 | 1010 | |||
| AK171091.1 | - | 6 | 1069 | |||
| BC004763.1 | - | 3 | 175 | |||
| BC012849.1 | - | 6 | 1148 | 3587716 | ||

Hnrnpll
Genomic Map