siRNA Details
| Gene Symbol: Zcchc12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028325.3 | NP_082601.1 | 5 | 1228 | ||
| GenBank mRNA | AK012833.1 | - | 5 | 1145 | ||
| AY466375.1 | - | 2 | 994 | |||
| BC049071.1 | - | 4 | 1124 | 5701407 | ||
| BC055714.1 | - | 4 | 1095 | 6415226 | ||

Zcchc12
Genomic Map