siRNA Details
| Gene Symbol: Zfp618 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028326.1 | NP_082602.1 | 4 | 464 | ||
| GenBank mRNA | AK012848.1 | - | 6 | 620 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp618| Gene Symbol: Zfp618 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028326.1 | NP_082602.1 | 4 | 464 | ||
| GenBank mRNA | AK012848.1 | - | 6 | 620 | ||