siRNA Details
| Gene Symbol: Zfp248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028335.2 | NP_082611.1 | 6 | 1571 | ||
| GenBank mRNA | AK012874.1 | - | 5 | 1522 | ||
| BC049087.1 | - | 3 | 1089 | |||
| BC082571.1 | - | 6 | 1571 | 30615600 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp248| Gene Symbol: Zfp248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028335.2 | NP_082611.1 | 6 | 1571 | ||
| GenBank mRNA | AK012874.1 | - | 5 | 1522 | ||
| BC049087.1 | - | 3 | 1089 | |||
| BC082571.1 | - | 6 | 1571 | 30615600 | ||