siRNA Details
| Gene Symbol: Zfp74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178384.3 | NP_848471.2 | 5 | 512 | ||
| GenBank mRNA | AK049278.1 | - | 448 | |||
| BC034883.1 | - | 5 | 536 | |||
| BC050048.1 | - | 5 | 512 | 30011303 | ||
| U29500.1 | - | 3 | 192 | |||

Zfp74
Genomic Map