siRNA Details
| Gene Symbol: Tmem161b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175187.5 | NP_780396.2 | 10 | 1084 | ||
| GenBank mRNA | AK013305.1 | - | 9 | 913 | ||
| AK088393.1 | - | 10 | 1056 | |||
| BC037611.1 | - | 8 | 864 | |||
| BC051663.1 | - | 10 | 1032 | 30020101 | ||
| BC060067.1 | - | 10 | 1126 | |||

Tmem161b
Genomic Map