siRNA Details
| Gene Symbol: MYBPH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004997.2 | NP_004988.2 | 3 | 556 | ||
| GenBank mRNA | BC044226.1 | - | 3 | 556 | 6176021 | |
| L05606.1 | - | 3 | 525 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MYBPH| Gene Symbol: MYBPH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004997.2 | NP_004988.2 | 3 | 556 | ||
| GenBank mRNA | BC044226.1 | - | 3 | 556 | 6176021 | |
| L05606.1 | - | 3 | 525 | |||