siRNA Details
| Gene Symbol: Ttc39c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028341.4 | NP_082617.2 | 4 | 744 | ||
| GenBank mRNA | AK013267.1 | - | 4 | 479 | ||
| BC020021.1 | - | 4 | 494 | 2648772 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttc39c| Gene Symbol: Ttc39c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028341.4 | NP_082617.2 | 4 | 744 | ||
| GenBank mRNA | AK013267.1 | - | 4 | 479 | ||
| BC020021.1 | - | 4 | 494 | 2648772 | ||