siRNA Details
| Gene Symbol: Arhgef10l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001112722.1 | NP_001106193.1 | 10, 11 | 1098 | ||
| NM_001112723.1 | NP_001106194.1 | 10, 11 | 1071 | |||
| NM_001290803.1 | NP_001277732.1 | 11, 12 | 1197 | |||
| NM_172415.3 | NP_766003.3 | 11, 12 | 1188 | |||
| GenBank mRNA | AK028570.1 | - | 4, 5 | 513 | ||
| AK028648.1 | - | 10, 11 | 1081 | |||
| AK129408.1 | - | 10, 11 | 1071 | |||
| BC034061.1 | - | 4, 5 | 334 | |||
| BC059220.1 | - | 11, 12 | 1176 | 6417641 | ||

Arhgef10l
Genomic Map