siRNA Details
| Gene Symbol: Tmem135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028343.4 | NP_082619.3 | 8 | 790 | ||
| GenBank mRNA | AK013269.1 | - | 8 | 764 | ||
| AK028401.1 | - | 8 | 600 | |||
| AK028846.1 | - | 8 | 764 | |||
| AK031818.1 | - | 4 | 209 | |||
| AK047200.1 | - | 8 | 764 | |||
| AY040841.1 | - | 8 | 640 | |||
| BC033279.1 | - | 8 | 770 | 4165990 | ||
| BC050931.1 | - | 8 | 699 | 6811187 | ||

Tmem135
Genomic Map