siRNA Details
Gene Symbol: Tmem135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028343.4 | NP_082619.3 | 13 | 1308 | ||
GenBank mRNA | AK013269.1 | - | 13 | 1282 | ||
AK028401.1 | - | 13 | 1118 | |||
AK028846.1 | - | 13 | 1282 | |||
AK031818.1 | - | 9 | 727 | |||
AK153175.1 | - | 13 | 1232 | |||
AY040841.1 | - | 13 | 1158 | |||
BC033279.1 | - | 13 | 1288 | 4165990 | ||
BC050931.1 | - | 13 | 1217 | 6811187 |

More Information
Other Species Targeted

Gene Ontology Categories:
