siRNA Details
| Gene Symbol: Fance | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163819.1 | NP_001157291.1 | 9, 10 | 1589 | ||
| GenBank mRNA | AK182233.1 | - | 2, 3 | 158 | ||
| AK184214.1 | - | 2, 3 | 157 | |||
| AK188566.1 | - | 2, 3 | 157 | |||
| AK207586.1 | - | 2, 3 | 157 | |||
| AK219855.1 | - | 2, 3 | 157 | |||
| BC039987.1 | - | 9, 10 | 1523 | |||

Fance
Genomic Map