siRNA Details
| Gene Symbol: Sass6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289568.1 | NP_001276497.1 | 8 | 855 | ||
| NM_001289571.1 | NP_001276500.1 | 6 | 711 | |||
| NM_028349.3 | NP_082625.2 | 7 | 772 | |||
| GenBank mRNA | AK013338.1 | - | 8 | 747 | ||
| AK145076.1 | - | 6 | 660 | |||
| AK165224.1 | - | 8 | 801 | |||
| BC050110.1 | - | 7 | 722 | 30024366 | ||

Sass6
Genomic Map