siRNA Details
| Gene Symbol: Zfp429 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080941.1 | NP_001074410.1 | 4 | 682 | ||
| GenBank mRNA | AK083314.1 | - | 5 | 750 | ||
| BC046484.1 | - | 4 | 666 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp429| Gene Symbol: Zfp429 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080941.1 | NP_001074410.1 | 4 | 682 | ||
| GenBank mRNA | AK083314.1 | - | 5 | 750 | ||
| BC046484.1 | - | 4 | 666 | |||