siRNA Details
Gene Symbol: Zfp429 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080941.1 | NP_001074410.1 | 4 | 1276 | ||
GenBank mRNA | AK083314.1 | - | 5 | 1345 | ||
BC046484.1 | - | 4 | 1260 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp429 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080941.1 | NP_001074410.1 | 4 | 1276 | ||
GenBank mRNA | AK083314.1 | - | 5 | 1345 | ||
BC046484.1 | - | 4 | 1260 |