siRNA Details
| Gene Symbol: Crtac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145123.4 | NP_660105.3 | 11 | 1546 | ||
| GenBank mRNA | AJ421516.1 | - | 11 | 1396 | ||
| AK032328.1 | - | 11 | 1411 | |||
| AK049801.1 | - | 11 | 1517 | |||
| AK083221.1 | - | 11 | 1550 | |||
| AK149020.1 | - | 11 | 1552 | |||
| AK149069.1 | - | 11 | 1547 | |||
| BC024472.1 | - | 8 | 982 | 4976509 | ||

Crtac1
Genomic Map