siRNA Details
| Gene Symbol: Kctd17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289671.1 | NP_001276600.1 | 3 | 461 | ||
| NM_001289672.1 | NP_001276601.1 | 3 | 461 | |||
| NM_001289673.1 | NP_001276602.1 | 3 | 461 | |||
| GenBank mRNA | AK179755.1 | - | 2 | 149 | ||
| BC144818.1 | - | 3 | 461 | |||
| BC144820.1 | - | 3 | 461 | |||
| BC145799.1 | - | 3 | 461 | 40131102 | ||

Kctd17
Genomic Map