siRNA Details
| Gene Symbol: Ccdc176 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028377.3 | NP_082653.2 | 2 | 212 | ||
| GenBank mRNA | AK013494.1 | - | 2 | 213 | ||
| AK133381.1 | - | 2 | 167 | |||
| BC151099.1 | - | 2 | 134 | 9088012 | ||
| BC151103.1 | - | 2 | 134 | 9088016 | ||

Ccdc176
Genomic Map