siRNA Details
| Gene Symbol: Xlr4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183094.3 | NP_898917.2 | 9 | 967 | ||
| GenBank mRNA | AK166871.1 | - | 9 | 968 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Xlr4c| Gene Symbol: Xlr4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183094.3 | NP_898917.2 | 9 | 967 | ||
| GenBank mRNA | AK166871.1 | - | 9 | 968 | ||