siRNA Details
| Gene Symbol: Setd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028385.1 | NP_082661.1 | 10 | 1356 | ||
| GenBank mRNA | AK013649.1 | - | 6 | 1286 | ||
| AK049143.1 | - | 4 | 434 | |||
| AK122551.1 | - | 10 | 1356 | |||
| AK164036.1 | - | 5 | 3261 | |||
| BC056502.1 | - | 12 | 1892 | |||
| BC083184.1 | - | 9 | 1237 | 30536831 | ||

Setd5
Genomic Map