siRNA Details
Gene Symbol: Fbxo47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081435.1 | NP_001074904.1 | 11 | 1666 | ||
GenBank mRNA | AK013672.1 | - | 2 | 368 | ||
BC099560.1 | - | 2 | 362 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fbxo47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081435.1 | NP_001074904.1 | 11 | 1666 | ||
GenBank mRNA | AK013672.1 | - | 2 | 368 | ||
BC099560.1 | - | 2 | 362 |