siRNA Details
| Gene Symbol: Cnih3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160211.1 | NP_001153683.1 | 1, 2 | 883 | ||
| NM_001160212.1 | NP_001153684.1 | 1, 2 | 883 | |||
| NM_028408.3 | NP_082684.1 | 1, 2 | 883 | |||
| GenBank mRNA | AK013789.1 | - | 1, 2 | 880 | ||
| AK033911.1 | - | 1, 2 | 881 | |||
| BC115640.1 | - | 1, 2 | 509 | 40103001 | ||
| BC115641.1 | - | 1, 2 | 509 | 40103002 | ||
| BG800984.1 | - | 1, 2 | 313 | |||

Cnih3
Genomic Map