siRNA Details
| Gene Symbol: Prkrir | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028410.1 | NP_082686.1 | 5 | 1727 | ||
| GenBank mRNA | AK013663.1 | - | 5 | 1818 | ||
| AK028175.1 | - | 1 | 832 | |||
| AK217173.1 | - | 1 | 41 | |||
| BC049103.1 | - | 5 | 1727 | 6400126 | ||

Prkrir
Genomic Map