siRNA Details
| Gene Symbol: Tmem192 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028427.3 | NP_082703.1 | 4, 5 | 695 | ||
| GenBank mRNA | AK013998.1 | - | 4, 5 | 694 | ||
| AK046181.1 | - | 4, 5 | 667 | |||
| AK132619.1 | - | 4, 5 | 666 | |||
| AK168146.1 | - | 4, 5 | 678 | |||

Tmem192
Genomic Map