siRNA Details
| Gene Symbol: Ppil6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028430.1 | NP_082706.1 | 1 | 162 | ||
| GenBank mRNA | AK013818.1 | - | 1 | 125 | ||
| BC147756.1 | - | 1 | 158 | 9056705 | ||
| BC147757.1 | - | 1 | 158 | 9056706 | ||
| BY104076.1 | - | 1 | 163 | |||
| CA466046.1 | - | 1 | 144 | |||

Ppil6
Genomic Map