siRNA Details
| Gene Symbol: Rps6ka5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153587.2 | NP_705815.1 | 9 | 1164 | ||
| GenBank mRNA | AK030051.1 | - | 8 | 1105 | ||
| AK046319.1 | - | 9 | 941 | |||
| AK160758.1 | - | 2 | 123 | |||
| AY341873.1 | - | 10 | 1153 | |||
| AY341881.1 | - | 9 | 1153 | |||
| BC035298.1 | - | 10 | 1164 | 4021865 | ||

Rps6ka5
Genomic Map