siRNA Details
| Gene Symbol: Fam101a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028443.2 | NP_082719.1 | 3 | 414 | ||
| GenBank mRNA | AK014112.1 | - | 3 | 444 | ||
| BC053444.1 | - | 3 | 414 | 6336076 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam101a| Gene Symbol: Fam101a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028443.2 | NP_082719.1 | 3 | 414 | ||
| GenBank mRNA | AK014112.1 | - | 3 | 444 | ||
| BC053444.1 | - | 3 | 414 | 6336076 | ||