siRNA Details
| Gene Symbol: Prrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028447.3 | NP_082723.2 | 3 | 567 | ||
| GenBank mRNA | AK004443.1 | - | 3 | 562 | ||
| AK143537.1 | - | 3 | 559 | |||
| AK179465.1 | - | 1 | 24 | |||
| BC014817.1 | - | 3 | 483 | 3709357 | ||

Prrc1
Genomic Map