siRNA Details
| Gene Symbol: Clec4a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204241.1 | NP_001191170.1 | 6 | 576 | ||
| NM_153197.4 | NP_694877.2 | 5 | 712 | |||
| GenBank mRNA | AK014135.1 | - | 5 | 611 | ||
| AK156040.1 | - | 5 | 712 | |||
| AY397674.1 | - | 5 | 523 | |||
| BC034893.1 | - | 6 | 576 | 1496162 | ||

Clec4a3
Genomic Map