siRNA Details
| Gene Symbol: Fezf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028462.1 | NP_082738.1 | 1, 2 | 995 | ||
| GenBank mRNA | AK014242.1 | - | 1, 2 | 873 | ||
| BC049157.1 | - | 1, 2 | 1094 | |||
| BC119565.1 | - | 1, 2 | 995 | 40057718 | ||
| BC119566.1 | - | 1, 2 | 995 | 40057721 | ||

Fezf1
Genomic Map